siRNA Details
| Gene Symbol: DNAJC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022365.3 | NP_071760.2 | 4, 5 | 827 | ||
| XM_011519614.2 | XP_011517916.1 | 4, 5 | 874 | |||
| XM_017016536.1 | XP_016872025.1 | 4, 5 | 874 | |||
| GenBank mRNA | AK026062.1 | - | 1, 2 | 101 | ||
| AK027263.1 | - | 4, 5 | 647 | |||
| AK308369.1 | - | 4, 5 | 833 | |||
| AY225122.1 | - | 4, 5 | 540 | |||
| BC110894.1 | - | 4, 5 | 827 | 5164559 | ||

DNAJC1
Genomic Map