siRNA Details
| Gene Symbol: ROBO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022370.3 | NP_071765.2 | 20, 21 | 3147 | ||
| XM_017018122.1 | XP_016873611.1 | 19, 20 | 2912 | |||
| GenBank mRNA | AK291045.1 | - | 20, 21 | 3081 | ||
| AK298792.1 | - | 3, 4 | 247 | |||
| AY509035.1 | - | 20, 21 | 3170 | |||
| BC008623.1 | - | 1, 2 | 263 | |||

ROBO3
Genomic Map