siRNA Details
| Gene Symbol: NOC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022451.10 | NP_071896.8 | 7 | 868 | ||
| XM_017016544.1 | XP_016872033.1 | 5 | 921 | |||
| GenBank mRNA | AB077992.1 | - | 7 | 755 | ||
| AK091246.1 | - | 7 | 825 | |||
| AK292520.1 | - | 7 | 850 | |||
| BC049850.1 | - | 7 | 818 | 5549517 | ||

NOC3L
Genomic Map