siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 15, 16 | 2072 | ||
| XM_011545916.2 | XP_011544218.1 | 14, 15 | 3419 | |||
| XM_011545917.2 | XP_011544219.1 | 14, 15 | 3419 | |||
| XM_011545919.2 | XP_011544221.1 | 13, 14 | 3382 | |||
| GenBank mRNA | AK021680.1 | - | 3, 4 | 263 | ||
| AK022551.1 | - | 6, 7 | 477 | |||
| AK096322.1 | - | 7, 8 | 855 | |||
| BQ276414.1 | - | 8, 9 | 640 | |||
| DA527487.1 | - | 7, 8 | 496 | |||
| DR761499.1 | - | 8, 9 | 569 | |||

FBRS
Genomic Map