siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 14 | 1895 | ||
| XM_011545916.2 | XP_011544218.1 | 13 | 3242 | |||
| XM_011545917.2 | XP_011544219.1 | 13 | 3242 | |||
| XM_011545919.2 | XP_011544221.1 | 12 | 3205 | |||
| GenBank mRNA | AK021680.1 | - | 2 | 86 | ||
| AK022551.1 | - | 5 | 300 | |||
| AK096322.1 | - | 6 | 678 | |||
| AK298229.1 | - | 7 | 458 | |||
| BQ276414.1 | - | 7 | 463 | |||
| DA527487.1 | - | 6 | 319 | |||
| DR761499.1 | - | 7 | 392 | |||

FBRS
Genomic Map