siRNA Details
| Gene Symbol: CPT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031847.2 | NP_001027017.1 | 10 | 1170 | ||
| NM_001876.3 | NP_001867.2 | 10 | 1170 | |||
| XM_005273762.2 | XP_005273819.1 | 10 | 1146 | |||
| XM_005273763.1 | XP_005273820.1 | 10 | 1146 | |||
| XM_017017220.1 | XP_016872709.1 | 10 | 1279 | |||
| GenBank mRNA | AK025307.1 | - | 5 | 456 | ||
| AK172798.1 | - | 10 | 1156 | |||
| AK309464.1 | - | 10 | 1146 | |||
| AK314301.1 | - | 10 | 1109 | |||
| BC000185.1 | - | 10 | 1045 | 3352642 | ||
| BT009791.1 | - | 9 | 1000 | |||
| L39211.1 | - | 10 | 1076 | |||

CPT1A
Genomic Map