siRNA Details
| Gene Symbol: IKZF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022465.3 | NP_071910.3 | 6 | 1430 | ||
| XM_005269086.3 | XP_005269143.1 | 8 | 972 | |||
| XM_005269089.2 | XP_005269146.1 | 6 | 1074 | |||
| XM_005269090.3 | XP_005269147.1 | 7 | 878 | |||
| XM_005269093.2 | XP_005269150.1 | 5 | 1349 | |||
| XM_011538664.1 | XP_011536966.1 | 8 | 975 | |||
| XM_011538666.2 | XP_011536968.1 | 7 | 823 | |||
| XM_011538669.2 | XP_011536971.1 | 6 | 773 | |||
| XM_017019805.1 | XP_016875294.1 | 10 | 1220 | |||
| XM_017019806.1 | XP_016875295.1 | 10 | 1463 | |||
| XM_017019807.1 | XP_016875296.1 | 10 | 1460 | |||
| XM_017019808.1 | XP_016875297.1 | 9 | 1145 | |||
| XM_017019809.1 | XP_016875298.1 | 10 | 1217 | |||
| XM_017019810.1 | XP_016875299.1 | 10 | 1315 | |||
| XM_017019811.1 | XP_016875300.1 | 9 | 1142 | |||
| XM_017019812.1 | XP_016875301.1 | 8 | 1218 | |||
| XM_017019813.1 | XP_016875302.1 | 8 | 1215 | |||
| XM_017019814.1 | XP_016875303.1 | 7 | 1124 | |||
| XM_017019815.1 | XP_016875304.1 | 7 | 1121 | |||
| XM_017019816.1 | XP_016875305.1 | 7 | 881 | |||
| GenBank mRNA | AB058685.1 | - | 4 | 974 | ||
| AF230809.1 | - | 5 | 682 | |||
| AK122880.1 | - | 5 | 1197 | |||
| AK314543.1 | - | 5 | 795 | |||
| BX647761.1 | - | 7 | 1000 | |||

IKZF4
Genomic Map