siRNA Details
| Gene Symbol: HHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022475.2 | NP_071920.1 | 6 | 1684 | ||
| XM_005263178.4 | XP_005263235.1 | 6 | 1553 | |||
| XM_006714288.3 | XP_006714351.1 | 6 | 1553 | |||
| GenBank mRNA | AK291849.1 | - | 6 | 1024 | ||
| AY009317.1 | - | 6 | 1524 | |||
| AY009951.1 | - | 6 | 1659 | |||
| AY358747.1 | - | 6 | 1634 | |||
| BC025311.1 | - | 6 | 1510 | 4122618 | ||

HHIP
Genomic Map