siRNA Details
| Gene Symbol: AKTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012398.2 | NP_001012398.1 | 3 | 363 | ||
| NM_001308325.1 | NP_001295254.1 | 3 | 334 | |||
| NM_022476.3 | NP_071921.1 | 3 | 334 | |||
| XM_005256095.4 | XP_005256152.1 | 3 | 683 | |||
| XM_005256096.4 | XP_005256153.1 | 3 | 261 | |||
| XM_005256097.4 | XP_005256154.1 | 3 | 295 | |||
| XM_005256098.4 | XP_005256155.1 | 3 | 259 | |||
| XM_017023564.1 | XP_016879053.1 | 3 | 683 | |||
| XM_017023565.1 | XP_016879054.1 | 3 | 261 | |||
| XM_017023566.1 | XP_016879055.1 | 3 | 259 | |||
| GenBank mRNA | AK023320.1 | - | 3 | 269 | ||
| AK222743.1 | - | 3 | 266 | |||
| AK303503.1 | - | 3 | 236 | |||
| BC001134.2 | - | 3 | 282 | 2967019 | ||
| BC095401.1 | - | 3 | 256 | 30334599 | ||
| CR457308.1 | - | 2 | 106 | |||

AKTIP
Genomic Map