siRNA Details
| Gene Symbol: CDH24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022478.3 | NP_071923.2 | 13 | 2304 | ||
| NM_144985.3 | NP_659422.2 | 12 | 2190 | |||
| GenBank mRNA | AK025342.1 | - | 5 | 608 | ||
| AK289576.1 | - | 12 | 2190 | |||
| AL137477.1 | - | 4 | 763 | |||
| AY260900.1 | - | 11 | 2023 | |||
| AY260901.1 | - | 12 | 2137 | |||
| AY358199.1 | - | 12 | 2146 | |||
| BC094810.1 | - | 1 | 68 | |||
| BC110867.1 | - | 5 | 760 | |||

CDH24
Genomic Map