siRNA Details
| Gene Symbol: PCNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022495.5 | NP_071940.4 | 7 | 1812 | ||
| XM_005268009.2 | XP_005268066.1 | 8 | 3189 | |||
| GenBank mRNA | AK023008.1 | - | 2 | 300 | ||
| AK095489.1 | - | 7 | 1796 | |||
| BX648440.1 | - | 3 | 536 | |||

PCNX4
Genomic Map