siRNA Details
| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033225.5 | NP_150094.5 | 29 | 4984 | ||
| XM_011534752.2 | XP_011533054.1 | 29 | 5213 | |||
| XM_011534753.2 | XP_011533055.1 | 12 | 2091 | |||
| XM_011534754.1 | XP_011533056.1 | 5 | 827 | |||
| XM_017013731.1 | XP_016869220.1 | 29 | 5213 | |||
| GenBank mRNA | AB067477.1 | - | 10 | 1702 | ||
| AB209502.1 | - | 17 | 3036 | |||
| AF333704.2 | - | 30 | 4987 | |||
| AK126936.1 | - | 5 | 827 | |||
| AY017307.1 | - | 30 | 4882 | |||
| AY358174.1 | - | 13 | 2127 | |||

CSMD1
Genomic Map