siRNA Details
| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033225.5 | NP_150094.5 | 39 | 6275 | ||
| XM_011534752.2 | XP_011533054.1 | 39 | 6504 | |||
| XM_011534753.2 | XP_011533055.1 | 22 | 3382 | |||
| XM_011534754.1 | XP_011533056.1 | 15 | 2118 | |||
| XM_017013731.1 | XP_016869220.1 | 39 | 6504 | |||
| GenBank mRNA | AB067477.1 | - | 20 | 2993 | ||
| AB209502.1 | - | 27 | 4327 | |||
| AF333704.2 | - | 40 | 6281 | |||
| AK126936.1 | - | 15 | 2118 | |||
| AY017307.1 | - | 40 | 6176 | |||
| AY358174.1 | - | 23 | 3418 | |||

CSMD1
Genomic Map