siRNA Details
| Gene Symbol: MOSPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040097.1 | NP_001035186.1 | 4 | 548 | ||
| NM_001040098.1 | NP_001035187.1 | 4 | 464 | |||
| NM_001040099.1 | NP_001035188.1 | 3 | 617 | |||
| NM_023948.4 | NP_076438.1 | 3 | 647 | |||
| XM_005250529.4 | XP_005250586.1 | 4 | 469 | |||
| XM_005250531.3 | XP_005250588.1 | 4 | 494 | |||
| GenBank mRNA | BC005042.1 | - | 3 | 576 | 3689026 | |
| BC011653.2 | - | 3 | 628 | 4299899 | ||
| BC028475.2 | - | 3 | 552 | |||
| BQ923161.1 | - | 522 | ||||

MOSPD3
Genomic Map