siRNA Details
| Gene Symbol: PLA2G2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022819.3 | NP_073730.3 | 1, 2 | 222 | ||
| XM_011541955.1 | XP_011540257.1 | 1, 2 | 215 | |||
| XM_011541956.1 | XP_011540258.1 | 1, 2 | 226 | |||
| GenBank mRNA | AK093645.1 | - | 1, 2 | 226 | ||
| BX647458.1 | - | 1, 2 | 222 | |||

PLA2G2F
Genomic Map