siRNA Details
| Gene Symbol: EBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022659.3 | NP_073150.2 | 10 | 1216 | ||
| GenBank mRNA | AK021562.1 | - | 3 | 255 | ||
| AK304372.1 | - | 9 | 864 | |||
| AY700779.1 | - | 2 | 144 | |||
| BC069665.1 | - | 3 | 233 | |||
| BC069726.1 | - | 3 | 236 | |||
| BC069747.1 | - | 3 | 235 | |||
| BC069768.1 | - | 3 | 236 | |||
| BC074794.2 | - | 2 | 172 | |||
| BC113478.1 | - | 2 | 200 | |||
| BC113504.1 | - | 2 | 200 | |||

EBF2
Genomic Map