siRNA Details
| Gene Symbol: MFSD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033055.2 | NP_149044.2 | 3 | 230 | ||
| XM_005271118.1 | XP_005271175.1 | 4 | 506 | |||
| XM_011541957.1 | XP_011540259.1 | 3 | 385 | |||
| XM_017002084.1 | XP_016857573.1 | 4 | 473 | |||
| GenBank mRNA | AF427492.1 | - | 3 | 230 | ||
| AK057172.1 | - | 3 | 366 | |||

MFSD14A
Genomic Map