siRNA Details
| Gene Symbol: CSRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320559.1 | NP_001307488.1 | 5 | 1633 | ||
| NM_001320560.1 | NP_001307489.1 | 5 | 1434 | |||
| NM_033027.3 | NP_149016.2 | 5 | 1502 | |||
| XM_017007048.1 | XP_016862537.1 | 5 | 4598 | |||
| XM_017007049.1 | XP_016862538.1 | 3 | 886 | |||
| GenBank mRNA | AB053121.1 | - | 5 | 1479 | ||
| AB063301.1 | - | 5 | 1394 | |||
| BC028213.1 | - | 3 | 262 | |||
| BC038949.1 | - | 5 | 1449 | 5547938 | ||

CSRNP1
Genomic Map