siRNA Details
| Gene Symbol: CSRNP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320559.1 | NP_001307488.1 | 5 | 1527 | ||
| NM_001320560.1 | NP_001307489.1 | 5 | 1328 | |||
| NM_033027.3 | NP_149016.2 | 5 | 1396 | |||
| XM_017007048.1 | XP_016862537.1 | 5 | 4492 | |||
| XM_017007049.1 | XP_016862538.1 | 3 | 780 | |||
| GenBank mRNA | AB053121.1 | - | 5 | 1373 | ||
| AB063301.1 | - | 5 | 1288 | |||
| BC038949.1 | - | 5 | 1343 | 5547938 | ||

CSRNP1
Genomic Map