siRNA Details
| Gene Symbol: WDR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166426.1 | NP_001159898.1 | 3 | 1310 | ||
| NM_017883.4 | NP_060353.2 | 4 | 975 | |||
| NR_029427.1 | 4 | 941 | ||||
| XM_006724539.1 | XP_006724602.1 | 5 | 628 | |||
| XM_006724541.3 | XP_006724604.1 | 5 | 594 | |||
| XM_011543947.2 | XP_011542249.1 | 5 | 588 | |||
| GenBank mRNA | AF158978.1 | - | 3 | 1337 | ||
| AF329819.4 | - | 4 | 975 | |||
| AK301186.1 | - | 2 | 161 | |||
| AK314739.1 | - | 4 | 938 | |||
| BC002507.2 | - | 1 | 37 | |||
| BC067094.1 | - | 4 | 622 | |||
| BC080579.1 | - | 4 | 704 | 6644248 | ||
| BI554303.1 | - | 4 | 685 | |||
| DQ907252.1 | - | 5 | 667 | |||
| DQ907253.1 | - | 3 | 982 | |||
| DQ907254.1 | - | 3 | 976 | |||
| DQ907255.1 | - | 3 | 925 | |||
| L08237.1 | - | 2 | 1996 | |||

WDR13
Genomic Map