siRNA Details
| Gene Symbol: WDR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166426.1 | NP_001159898.1 | 1 | 1108 | ||
| NM_017883.4 | NP_060353.2 | 2 | 773 | |||
| NR_029427.1 | 2 | 739 | ||||
| XM_006724539.1 | XP_006724602.1 | 3 | 426 | |||
| XM_006724541.3 | XP_006724604.1 | 3 | 392 | |||
| XM_011543947.2 | XP_011542249.1 | 3 | 386 | |||
| GenBank mRNA | AF158978.1 | - | 1 | 1135 | ||
| AF329819.4 | - | 2 | 773 | |||
| BC067094.1 | - | 2 | 420 | |||
| BC080579.1 | - | 2 | 502 | 6644248 | ||
| BI554303.1 | - | 2 | 478 | |||
| DQ907252.1 | - | 3 | 465 | |||
| DQ907253.1 | - | 1 | 780 | |||
| DQ907254.1 | - | 1 | 773 | |||
| DQ907255.1 | - | 1 | 723 | |||
| L08237.1 | - | 1 | 1416 | |||

WDR13
Genomic Map