siRNA Details
| Gene Symbol: WDR13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166426.1 | NP_001159898.1 | 5 | 1715 | ||
| NM_017883.4 | NP_060353.2 | 6 | 1380 | |||
| NR_029427.1 | 6 | 1346 | ||||
| XM_006724539.1 | XP_006724602.1 | 7 | 1033 | |||
| XM_006724541.3 | XP_006724604.1 | 7 | 999 | |||
| XM_011543947.2 | XP_011542249.1 | 7 | 993 | |||
| GenBank mRNA | AF158978.1 | - | 5 | 1742 | ||
| AF329819.4 | - | 6 | 1380 | |||
| AK000570.1 | - | 1 | 26 | |||
| AK225542.1 | - | 1 | 26 | |||
| AK314739.1 | - | 6 | 1343 | |||
| BC002507.2 | - | 3 | 442 | |||
| BC067094.1 | - | 6 | 1027 | |||
| BC080579.1 | - | 6 | 1109 | 6644248 | ||
| BX649101.1 | - | 4 | 325 | |||
| DQ907252.1 | - | 7 | 1072 | |||
| DQ907253.1 | - | 5 | 1387 | |||
| DQ907255.1 | - | 5 | 1330 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028542, NM_026137, NM_001290783, AK006837, AK017549, and others.

WDR13
Genomic Map