siRNA Details
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022735.3 | NP_073572.2 | 7 | 1180 | ||
| GenBank mRNA | AB043587.2 | - | 7 | 1188 | ||
| AK025520.1 | - | 7 | 1181 | |||
| AK314468.1 | - | 7 | 1181 | |||
| AY150218.1 | - | 7 | 1159 | |||
| BC034563.2 | - | 5 | 629 | |||
| BC045533.1 | - | 7 | 1166 | 5259930 | ||
| BC060792.1 | - | 7 | 1176 | 4813993 | ||

ACBD3
Genomic Map