siRNA Details
| Gene Symbol: MFSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289407.1 | NP_001276336.1 | 2, 3 | 405 | ||
| NM_022736.2 | NP_073573.2 | 2, 3 | 436 | |||
| GenBank mRNA | AB014732.1 | - | 2, 3 | 268 | ||
| AK024215.1 | - | 2, 3 | 239 | |||
| AK297034.1 | - | 255 | ||||
| AK297593.1 | - | 2, 3 | 436 | |||
| AK301680.1 | - | 2, 3 | 175 | |||
| BC042197.1 | - | 2, 3 | 248 | 5737259 | ||

MFSD1
Genomic Map