siRNA Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170635.1 | NP_001164106.1 | 3, 4 | 451 | ||
| NM_022737.2 | NP_073574.2 | 4 | 526 | |||
| XM_005260017.1 | XP_005260074.1 | 4 | 526 | |||
| XM_017027147.1 | XP_016882636.1 | 4 | 529 | |||
| XM_017027148.1 | XP_016882637.1 | 3, 4 | 447 | |||
| GenBank mRNA | AK023117.1 | - | 4 | 487 | ||
| AK094869.1 | - | 3, 4 | 339 | |||
| AK225482.1 | - | 4 | 441 | |||
| AY304516.1 | - | 4 | 481 | |||
| AY339628.1 | - | 3 | 181 | |||
| BC009378.1 | - | 4 | 369 | 4125316 | ||

PLPPR2
Genomic Map