siRNA Details
Gene Symbol: CCDC136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201372.1 | NP_001188301.1 | 3 | 363 | ||
NM_022742.4 | NP_073579.4 | 2 | 545 | |||
XM_005250534.3 | XP_005250591.1 | 3 | 329 | |||
XM_005250538.1 | XP_005250595.1 | 3 | 329 | |||
XM_011516483.1 | XP_011514785.1 | 3 | 329 | |||
XM_011516484.1 | XP_011514786.1 | 3 | 329 | |||
XM_011516485.1 | XP_011514787.1 | 3 | 329 | |||
XM_011516486.1 | XP_011514788.1 | 3 | 329 | |||
XM_011516487.2 | XP_011514789.1 | 3 | 329 | |||
XM_011516488.2 | XP_011514790.1 | 3 | 329 | |||
XM_011516489.1 | XP_011514791.1 | 3 | 329 | |||
XM_011516490.2 | XP_011514792.1 | 3 | 329 | |||
XM_017012532.1 | XP_016868021.1 | 3 | 329 | |||
XM_017012533.1 | XP_016868022.1 | 3 | 795 | |||
XM_017012534.1 | XP_016868023.1 | 3 | 329 | |||
XM_017012535.1 | XP_016868024.1 | 3 | 329 | |||
XM_017012536.1 | XP_016868025.1 | 3 | 329 | |||
GenBank mRNA | AB058696.1 | - | 2 | 496 | ||
AK124447.1 | - | 3 | 329 | |||
BC150331.1 | - | 2 | 507 | 8860498 | ||
BC152408.1 | - | 2 | 496 | 8862511 | ||
DR003042.1 | - | 4 | 454 |
