siRNA Details
| Gene Symbol: FAM160B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022749.5 | NP_073586.5 | 6 | 593 | ||
| XM_005273602.3 | XP_005273659.1 | 6 | 593 | |||
| XM_006716381.2 | XP_006716444.1 | 6 | 593 | |||
| XM_006716385.3 | XP_006716448.1 | 7 | 2088 | |||
| XM_017013732.1 | XP_016869221.1 | 6 | 593 | |||
| XM_017013733.1 | XP_016869222.1 | 7 | 710 | |||
| XM_017013734.1 | XP_016869223.1 | 6 | 574 | |||
| GenBank mRNA | BC052237.1 | - | 6 | 590 | ||

FAM160B2
Genomic Map