siRNA Details
| Gene Symbol: PARP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022750.3 | NP_073587.1 | 6 | 2019 | ||
| NR_130117.1 | 6 | 2019 | ||||
| XM_005250038.3 | XP_005250095.1 | 6 | 2014 | |||
| XM_005250039.4 | XP_005250096.1 | 6 | 2016 | |||
| XM_005250040.4 | XP_005250097.1 | 6 | 2016 | |||
| XM_011516491.2 | XP_011514793.1 | 6 | 2014 | |||
| GenBank mRNA | AL136766.1 | - | 6 | 1641 | ||
| AL137255.1 | - | 6 | 838 | |||
| BC081541.1 | - | 6 | 2019 | 5296560 | ||

PARP12
Genomic Map