siRNA Details
| Gene Symbol: FNDC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135095.1 | NP_001128567.1 | 6 | 654 | ||
| NM_022763.3 | NP_073600.3 | 6 | 727 | |||
| XM_017007064.1 | XP_016862553.1 | 6 | 707 | |||
| GenBank mRNA | AB098597.1 | - | 5 | 612 | ||
| AK127826.1 | - | 6 | 707 | |||
| AK223599.1 | - | 6 | 758 | |||
| AK314478.1 | - | 6 | 654 | |||
| AY358367.1 | - | 6 | 696 | |||
| BC039297.1 | - | 6 | 618 | 5297602 | ||
| BX648340.1 | - | 6 | 649 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173182, BC067389, AK220297, AB098596, AK045099, and others.

FNDC3B
Genomic Map