siRNA Details
| Gene Symbol: AEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022767.3 | NP_073604.3 | 2 | 551 | ||
| XM_005254966.1 | XP_005255023.1 | 2 | 741 | |||
| XM_005254967.1 | XP_005255024.1 | 2 | 881 | |||
| XM_006720645.3 | XP_006720708.2 | 2 | 510 | |||
| XM_011521905.2 | XP_011520207.1 | 4 | 905 | |||
| XM_017022489.1 | XP_016877978.1 | 2 | 1528 | |||
| XM_017022490.1 | XP_016877979.1 | 4 | 868 | |||
| GenBank mRNA | AF318330.1 | - | 2 | 481 | ||
| AF327352.1 | - | 1 | 472 | |||
| AK022546.1 | - | 1 | 85 | |||
| AK308119.1 | - | 2 | 511 | |||
| BC005164.1 | - | 1 | 467 | 3531883 | ||
| BC014407.1 | - | 1 | 86 | 4558021 | ||
| BC020988.1 | - | 2 | 489 | 3852419 | ||

AEN
Genomic Map