siRNA Details
| Gene Symbol: FNDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022823.2 | NP_073734.1 | 6 | 1002 | ||
| GenBank mRNA | AK026015.1 | - | 6 | 962 | ||
| AY358136.1 | - | 6 | 720 | |||
| BC032725.1 | - | 5 | 633 | 5533159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FNDC4| Gene Symbol: FNDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022823.2 | NP_073734.1 | 6 | 1002 | ||
| GenBank mRNA | AK026015.1 | - | 6 | 962 | ||
| AY358136.1 | - | 6 | 720 | |||
| BC032725.1 | - | 5 | 633 | 5533159 | ||