siRNA Details
| Gene Symbol: YTHDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022828.3 | NP_073739.3 | 3 | 492 | ||
| XM_011543581.1 | XP_011541883.1 | 3 | 503 | |||
| XM_011543582.2 | XP_011541884.1 | 3 | 477 | |||
| XM_011543586.2 | XP_011541888.1 | 3 | 477 | |||
| XM_017009730.1 | XP_016865219.1 | 3 | 503 | |||
| GenBank mRNA | AK131450.1 | - | 2 | 185 | ||
| AK310855.1 | - | 3 | 477 | |||
| BC137285.1 | - | 3 | 442 | 9021282 | ||
| CA943109.1 | - | 3 | 476 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001163013, BC171951, BC138263.

YTHDC2
Genomic Map