siRNA Details
| Gene Symbol: YTHDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022828.3 | NP_073739.3 | 5 | 933 | ||
| XM_011543581.1 | XP_011541883.1 | 5 | 944 | |||
| XM_011543582.2 | XP_011541884.1 | 5 | 918 | |||
| XM_011543586.2 | XP_011541888.1 | 5 | 918 | |||
| XM_017009730.1 | XP_016865219.1 | 5 | 944 | |||
| XM_017009731.1 | XP_016865220.1 | 4 | 719 | |||
| XM_017009732.1 | XP_016865221.1 | 4 | 719 | |||
| XM_017009733.1 | XP_016865222.1 | 3 | 547 | |||
| XM_017009734.1 | XP_016865223.1 | 3 | 547 | |||
| XM_017009735.1 | XP_016865224.1 | 3 | 515 | |||
| GenBank mRNA | AK096486.1 | - | 3 | 536 | ||
| AK131450.1 | - | 4 | 626 | |||
| AK310855.1 | - | 5 | 918 | |||
| BC137285.1 | - | 5 | 883 | 9021282 | ||

YTHDC2
Genomic Map