siRNA Details
| Gene Symbol: FAM129B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035534.2 | NP_001030611.1 | 5, 6 | 620 | ||
| NM_022833.3 | NP_073744.2 | 5, 6 | 832 | |||
| XM_005252135.2 | XP_005252192.2 | 6, 7 | 679 | |||
| XM_011518925.1 | XP_011517227.1 | 6, 7 | 851 | |||
| GenBank mRNA | AB210016.1 | - | 5, 6 | 570 | ||
| AF151783.1 | - | 5, 6 | 602 | |||
| AF192911.1 | - | 5, 6 | 823 | |||
| AL137555.1 | - | 4, 5 | 440 | |||
| CN423640.1 | - | 5, 6 | 622 | |||

FAM129B
Genomic Map