siRNA Details
| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022909.3 | NP_075060.1 | 8 | 594 | ||
| GenBank mRNA | AB035124.1 | - | 8 | 507 | ||
| AK123761.1 | - | 7 | 512 | |||
| AK290746.1 | - | 8 | 577 | |||
| BC012024.1 | - | 8 | 570 | 4510607 | ||
| BC015355.1 | - | 8 | 576 | 3451767 | ||
| KU178758.1 | - | 8 | 507 | |||
| KU178759.1 | - | 8 | 466 | |||

CENPH
Genomic Map