siRNA Details
| Gene Symbol: MRPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182640.2 | NP_872578.1 | 5 | 536 | ||
| XM_011511644.2 | XP_011509946.1 | 4 | 376 | |||
| GenBank mRNA | AK001451.1 | - | 5 | 478 | ||
| AK130078.1 | - | 5 | 477 | |||
| AV729381.1 | - | 5 | 513 | |||
| BC036575.1 | - | 6 | 489 | |||
| BC047784.1 | - | 4 | 186 | |||
| BC057240.1 | - | 5 | 490 | 6495350 | ||
| DT217261.1 | - | 5 | 536 | |||

MRPS9
Genomic Map