siRNA Details
| Gene Symbol: MRPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032478.3 | NP_115867.2 | 3 | 534 | ||
| GenBank mRNA | AK313283.1 | - | 3 | 349 | ||
| BC013311.2 | - | 3 | 333 | 3546083 | ||
| BC034460.1 | - | 3 | 288 | |||
| DB289191.1 | - | 3 | 534 | |||

MRPL38
Genomic Map