siRNA Details
| Gene Symbol: MRPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016050.4 | NP_057134.1 | 2 | 276 | ||
| NM_170738.3 | NP_733934.1 | 2 | 198 | |||
| NM_170739.3 | NP_733935.1 | 2 | 276 | |||
| NR_120588.1 | 2 | 276 | ||||
| NR_120589.1 | 2 | 126 | ||||
| XM_017018135.1 | XP_016873624.1 | 2 | 210 | |||
| GenBank mRNA | AB049638.1 | - | 2 | 248 | ||
| AF151871.1 | - | 2 | 260 | |||
| AK127603.1 | - | 2 | 885 | |||
| AK223112.1 | - | 2 | 252 | |||
| AK290084.1 | - | 2 | 254 | |||
| BC005002.1 | - | 2 | 255 | 3542758 | ||
| BC108277.1 | - | 2 | 171 | 4416083 | ||
| BI193255.1 | - | 2 | 190 | |||
| BM554886.1 | - | 2 | 238 | |||
| BM755421.1 | - | 2 | 267 | |||
| BU597133.1 | - | 2 | 279 | |||

MRPL11
Genomic Map