siRNA Details
| Gene Symbol: TMEM237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044385.2 | NP_001037850.1 | 10, 11 | 1082 | ||
| NM_152388.3 | NP_689601.2 | 10, 11 | 1247 | |||
| GenBank mRNA | AB053301.1 | - | 10, 11 | 1249 | ||
| AK090601.1 | - | 10, 11 | 940 | |||
| AK303954.1 | - | 10, 11 | 1069 | |||
| AK304395.1 | - | 8, 9 | 771 | |||
| BC013730.1 | - | 2, 3 | 165 | |||
| BC029611.1 | - | 10, 11 | 995 | 5304046 | ||

TMEM237
Genomic Map