siRNA Details
| Gene Symbol: RPL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000971.3 | NP_000962.2 | 1 | 557 | ||
| XM_006716463.2 | XP_006716526.1 | 5, 6 | 1150 | |||
| GenBank mRNA | AK225246.1 | - | 1 | 556 | ||
| AK225254.1 | - | 1 | 555 | |||
| AK291119.1 | - | 5, 6 | 555 | |||
| AK313365.1 | - | 5, 6 | 555 | |||
| BC006095.1 | - | 1 | 545 | 2821364 | ||
| BC008850.2 | - | 1 | 582 | 4138113 | ||
| BC009599.1 | - | 1 | 557 | 4134557 | ||
| BC071671.1 | - | 1 | 538 | 4392472 | ||
| BC071894.1 | - | 1 | 542 | 2906310 | ||
| BC071895.1 | - | 1 | 540 | 6568654 | ||
| BC087837.1 | - | 1 | 541 | 6170371 | ||
| CR456773.1 | - | 1 | 535 | |||
| L16558.1 | - | 1 | 535 | |||
| X52967.1 | - | 5, 6 | 556 | |||
| X57958.1 | - | 1 | 572 | |||
| X57959.1 | - | 1 | 545 | |||

RPL7
Genomic Map