siRNA Details
| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023010.3 | NP_075386.1 | 9 | 1344 | ||
| NM_080632.2 | NP_542199.1 | 10 | 1383 | |||
| XM_017029737.1 | XP_016885226.1 | 10 | 1383 | |||
| XM_017029738.1 | XP_016885227.1 | 10 | 1383 | |||
| XM_017029739.1 | XP_016885228.1 | 9 | 1344 | |||
| XM_017029740.1 | XP_016885229.1 | 10 | 1383 | |||
| GenBank mRNA | AF318576.1 | - | 10 | 1303 | ||
| AK093728.1 | - | 9 | 1282 | |||
| AY013251.1 | - | 9 | 1314 | |||
| BC121017.1 | - | 9 | 1287 | 40117102 | ||
| BC121018.1 | - | 8 | 823 | |||

UPF3B
Genomic Map