siRNA Details
| Gene Symbol: PRAMEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023014.1 | NP_075390.1 | 3 | 611 | ||
| GenBank mRNA | AL049674.1 | - | 3 | 611 | ||
| BC075008.2 | - | 2 | 560 | 30915615 | ||
| BC075009.2 | - | 2 | 560 | 30915402 | ||
| Gene Symbol: PRAMEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023013.4 | NP_075389.2 | 3 | 627 | ||
| GenBank mRNA | AK125568.1 | - | 3 | 627 | ||
| AL049686.1 | - | 3 | 627 | |||
| BC144199.1 | - | 3 | 597 | 9052719 | ||
| Gene Symbol: PRAMEF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291380.1 | NP_001278309.1 | 3 | 627 | ||
| Gene Symbol: PRAMEF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024661.2 | NP_001019832.2 | 3 | 627 | ||
| Gene Symbol: PRAMEF34P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111947.1 | 3 | 627 | |||
| Gene Symbol: PRAMEF36P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111945.1 | 3 | 627 | |||

PRAMEF2
Genomic Map