siRNA Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023070.2 | NP_075558.2 | 5 | 1041 | ||
| XM_005271136.1 | XP_005271193.1 | 6 | 1127 | |||
| XM_005271137.2 | XP_005271194.1 | 6 | 1124 | |||
| XM_005271138.3 | XP_005271195.1 | 5 | 1075 | |||
| XM_005271139.3 | XP_005271196.1 | 5 | 1072 | |||
| XM_017002147.1 | XP_016857636.1 | 6 | 1077 | |||
| GenBank mRNA | AK091612.1 | - | 5 | 1395 | ||
| AL137241.1 | - | 5 | 1024 | |||
| BC017498.1 | - | 5 | 662 | 4576469 | ||

ZFP69B
Genomic Map