siRNA Details
| Gene Symbol: PLEKHA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019091.3 | NP_061964.3 | 7 | 1128 | ||
| XM_011511689.2 | XP_011509991.1 | 8 | 1237 | |||
| GenBank mRNA | AF286162.1 | - | 7 | 967 | ||
| AK000074.1 | - | 5 | 531 | |||
| AK074209.1 | - | 7 | 1103 | |||
| AK314459.1 | - | 7 | 1132 | |||
| BC044567.1 | - | 7 | 822 | 5262097 | ||
| DQ786259.1 | - | 4 | 445 | |||
| DQ786318.1 | - | 3 | 285 | |||

PLEKHA3
Genomic Map