siRNA Details
| Gene Symbol: ZNF747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305018.1 | NP_001291947.1 | 3 | 1299 | ||
| NM_001305019.1 | NP_001291948.1 | 3 | 1296 | |||
| NM_001305020.1 | NP_001291949.1 | 2 | 1436 | |||
| NM_023931.3 | NP_076420.1 | 2 | 1439 | |||
| GenBank mRNA | AK054567.1 | - | 2 | 1338 | ||
| AK292192.1 | - | 2 | 1150 | |||
| AK299308.1 | - | 3 | 1181 | |||
| AK316254.1 | - | 3 | 1224 | |||
| BC001361.2 | - | 2 | 1342 | 3050694 | ||

ZNF747
Genomic Map