siRNA Details
| Gene Symbol: FUNDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023934.3 | NP_076423.2 | 4 | 596 | ||
| GenBank mRNA | AF267862.1 | - | 1 | 626 | ||
| AF320778.1 | - | 4 | 559 | |||
| AJ272054.1 | - | 4 | 553 | |||
| AK091092.1 | - | 1 | 2662 | |||
| AK129832.1 | - | 715 | ||||
| AK313143.1 | - | 4 | 538 | |||
| AY032594.1 | - | 4 | 446 | |||
| BC000255.1 | - | 4 | 543 | 3355861 | ||
| BC072685.1 | - | 4 | 501 | 6502640 | ||
| BC108657.1 | - | 4 | 452 | 5752287 | ||

FUNDC2
Genomic Map