siRNA Details
| Gene Symbol: C11orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144936.1 | NP_001138408.1 | 5 | 3723 | ||
| GenBank mRNA | AK096306.1 | - | 1 | 622 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11orf95| Gene Symbol: C11orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144936.1 | NP_001138408.1 | 5 | 3723 | ||
| GenBank mRNA | AK096306.1 | - | 1 | 622 | ||