siRNA Details
Gene Symbol: TMEM108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136469.2 | NP_001129941.1 | 4 | 1489 | ||
NM_023943.3 | NP_076432.1 | 4 | 1493 | |||
XM_011513097.2 | XP_011511399.1 | 3 | 3495 | |||
XM_011513098.2 | XP_011511400.1 | 5 | 1588 | |||
XM_017007080.1 | XP_016862569.1 | 5 | 1645 | |||
GenBank mRNA | AB051477.1 | - | 3 | 1303 | ||
AL136578.1 | - | 1 | 845 | |||
AY358381.1 | - | 4 | 1375 | |||
BC000568.2 | - | 4 | 1372 | 3163990 |
