siRNA Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024026.4 | NP_076931.1 | 2 | 1033 | ||
| XM_017020740.1 | XP_016876229.1 | 2 | 667 | |||
| GenBank mRNA | BC023616.2 | - | 2 | 590 | 4645641 | |
| BC068492.1 | - | 2 | 600 | 4801279 | ||
| BG773624.1 | - | 2 | 624 | |||
| BI601227.1 | - | 2 | 576 | |||

MRPL57
Genomic Map