siRNA Details
| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024051.3 | NP_076956.1 | 3 | 448 | ||
| NR_037669.1 | 2 | 302 | ||||
| GenBank mRNA | AK021779.1 | - | 2 | 927 | ||
| AK315608.1 | - | 3 | 439 | |||
| BC000625.2 | - | 3 | 443 | 3346822 | ||
| BC013937.1 | - | 3 | 409 | 3997629 | ||
| BC019243.1 | - | 3 | 440 | 3828006 | ||
| BG527593.1 | - | 2 | 291 | |||

GGCT
Genomic Map